(S)-2-Amino-3-(4-(prop-2-yn-1-yloxy)phenyl)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Propargyloxy-L-phenylalanine

A721556-100MG A721556-1G A721556-250MG A721556-5G
76651-576EA 97 USD
76651-576 76651-578 76651-580 77749-470
(S)-2-Amino-3-(4-(prop-2-yn-1-yloxy)phenyl)propanoic acid 97%
(S)-2-Amino-3-(4-(prop-2-yn-1-yloxy)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MDL Number: MFCD21363200
CAS Number: 610794-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR