Methyl 5-fluoro-2-(2-methoxy-2-oxoethyl)benzoate 97%

Supplier: Ambeed
Warning

A721065-1G A721065-5G
76651-572EA 293.87 USD
76651-572 76651-574
Methyl 5-fluoro-2-(2-methoxy-2-oxoethyl)benzoate 97%
Methyl 5-fluoro-2-(2-methoxy-2-oxoethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₄
MW: 226.20 g/mol
MDL Number: MFCD31714491
CAS Number: 2326068-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR