Methyl 2-(4-bromo-2-nitrophenyl)-2-(hydroxyimino)acetate 95%

Supplier: Ambeed
Danger

A721057-1G A721057-5G
76651-568EA 41.38 USD
76651-568 76651-570
Methyl 2-(4-bromo-2-nitrophenyl)-2-(hydroxyimino)acetate 95%
Methyl 2-(4-bromo-2-nitrophenyl)-2-(hydroxyimino)acetate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₅
MW: 303.07 g/mol
MDL Number: MFCD28134474
CAS Number: 1628563-38-1
UN: 2926
ADR: 4.1,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR