2,7-Bis(piperidin-1-ylsulfonyl)-9H-fluoren-9-one oxime 99+%

Supplier: Ambeed
Danger

Synonyms: CIL56

A720858-10MG A720858-1MG A720858-25MG A720858-5MG
76651-554EA 52 USD
76651-554 76651-556 76651-558 76651-560
2,7-Bis(piperidin-1-ylsulfonyl)-9H-fluoren-9-one oxime 99+%
2,7-Bis(piperidin-1-ylsulfonyl)-9H-fluoren-9-one oxime

Precaución: for Research Use Only

Formula: C₂₃H₂₇N₃O₅S₂
MDL Number: MFCD00323348
CAS Number: 300802-28-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR