5-(11bR)-Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl-5H-dibenz[b,f]azepine ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A720059-100MG
A720059-50MG
76651-540EA
39.26
USD
76651-540
76651-542
5-(11bR)-Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl-5H-dibenz[b,f]azepine ≥97%, ee 99%
5-(11bR)-Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl-5H-dibenz[b,f]azepine
Precaución: for Research Use Only
Formula:
C₃₄H₂₂NO₂P MW: 507.52 g/mol |
CAS Number:
1265884-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...