6-(Benzyloxy)-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A719054-1G A719054-250MG
76651-524EA 281.14 USD
76651-524 76651-526
6-(Benzyloxy)-1H-pyrrolo[2,3-b]pyridine 95%
6-(Benzyloxy)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O
MW: 224.2500 g/mol
MDL Number: MFCD16657185
CAS Number: 1018441-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR