8-Chloro-5-methyl-1-((1r,4r)-4-(pyridin-2-yloxy)cyclohexyl)-5,6-dihydro-4H-benzo[f][1,2,4]triazolo[4,3-a][1,4]diazepine 97%

Supplier: Ambeed
Warning

Synonyms: Balovaptan

A718871-10MG A718871-1MG A718871-25MG A718871-50MG
76651-510EA 94.43 USD
76651-510 76651-512 76651-514 76651-516
8-Chloro-5-methyl-1-((1r,4r)-4-(pyridin-2-yloxy)cyclohexyl)-5,6-dihydro-4H-benzo[f][1,2,4]triazolo[4,3-a][1,4]diazepine 97%
8-Chloro-5-methyl-1-((1r,4r)-4-(pyridin-2-yloxy)cyclohexyl)-5,6-dihydro-4H-benzo[f][1,2,4]triazolo[4,3-a][1,4]diazepine

Precaución: for Research Use Only

Formula: C₂₂H₂₄ClN₅O
MW: 409.91 g/mol
MDL Number: MFCD31706224
CAS Number: 1228088-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR