3-(4-(Trifluoromethyl)benzyl)azetidine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A718084-100MG
A718084-250MG
76651-504EA
126
USD
76651-504
76651-506
3-(4-(Trifluoromethyl)benzyl)azetidine hydrochloride 97%
3-(4-(Trifluoromethyl)benzyl)azetidine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₃ClF₃N |
MDL Number:
MFCD14705733 CAS Number: 1203685-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...