1-Thio-β-D-glucose sodium salt hydrate 96%

Supplier: Ambeed
Warning

A717314-100MG A717314-250MG
76651-482EA 101 USD
76651-482 76651-484
1-Thio-β-D-glucose sodium salt hydrate 96%
1-Thio-beta-D-glucose sodium salt hydrate

Precaución: for Research Use Only

Formula: C₆H₁₃NaO₆S
MDL Number: MFCD00067685
CAS Number: 255818-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR