Ethyl 5-(((2,4,6-trimethyl-N-((3'-(methylsulfonyl)-[1,1'-biphenyl]-4-yl)methyl)phenyl)sulfonamido)methyl)furan-2-carboxylate 98%

Supplier: Ambeed
Warning

A717181-10MG A717181-1MG A717181-25MG A717181-50MG A717181-5MG
76651-472EA 106 USD
76651-472 76651-474 76651-476 76651-478 76651-480
Ethyl 5-(((2,4,6-trimethyl-N-((3'-(methylsulfonyl)-[1,1'-biphenyl]-4-yl)methyl)phenyl)sulfonamido)methyl)furan-2-carboxylate 98%
Ethyl 5-(((2,4,6-trimethyl-N-((3'-(methylsulfonyl)-[1,1'-biphenyl]-4-yl)methyl)phenyl)sulfonamido)methyl)furan-2-carboxylate

Precaución: for Research Use Only

Formula: C₃₁H₃₃NO₇S₂
MDL Number: MFCD29472591
CAS Number: 1416153-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR