1-Phenylpropane-1,3-diol 97%

Supplier: Ambeed
Warning

A717097-1G A717097-250MG
76651-470EA 149.59 USD
76651-470 77617-948
1-Phenylpropane-1,3-diol 97%
1-Phenylpropane-1,3-diol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.1904 g/mol
MDL Number: MFCD00033357
CAS Number: 4850-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR