3-Cyclopropyl-4-methyl-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Danger

A716468-100MG A716468-1G A716468-250MG
76651-464EA 95.49 USD
76651-464 76651-466 76651-468
3-Cyclopropyl-4-methyl-1H-pyrazole-5-carboxylic acid 95%
3-Cyclopropyl-4-methyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.1700 g/mol
MDL Number: MFCD20535329
CAS Number: 1867157-28-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR