2,4,8-Trichloroquinazoline 95%

Supplier: Ambeed
Warning

A1349704-100MG A1349704-1G A1349704-5G
76658-130EA 32.89 USD
76658-130 77596-808 77621-628
2,4,8-Trichloroquinazoline 95%
2,4,8-Trichloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₃Cl₃N₂
MW: 233.482 g/mol
MDL Number: MFCD09954870
CAS Number: 62484-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR