N-(Prop-2-yn-1-yl)acrylamide 95%

Supplier: Ambeed
Warning

A715917-100MG A715917-1G A715917-250MG
76651-450EA 70 USD
76651-450 76651-452 76651-454
N-(Prop-2-yn-1-yl)acrylamide 95%
N-(Prop-2-yn-1-yl)acrylamide

Precaución: for Research Use Only

Formula: C₆H₇NO
MDL Number: MFCD12091202
CAS Number: 123523-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR