(E)-4-((2-(4-((E)-1-(1H-Indazol-5-yl)-2-phenylbut-1-en-1-yl)phenoxy)ethyl)amino)-N,N-dimethylbut-2-enamide 98%

Supplier: Ambeed
Warning

A715866-10MG A715866-1MG A715866-25MG A715866-5MG
76651-442EA 119.89 USD
76651-442 76651-444 76651-446 76651-448
(E)-4-((2-(4-((E)-1-(1H-Indazol-5-yl)-2-phenylbut-1-en-1-yl)phenoxy)ethyl)amino)-N,N-dimethylbut-2-enamide 98%
(E)-4-((2-(4-((E)-1-(1H-Indazol-5-yl)-2-phenylbut-1-en-1-yl)phenoxy)ethyl)amino)-N,N-dimethylbut-2-enamide

Precaución: for Research Use Only

Formula: C₃₁H₃₄N₄O₂
MW: 494.63 g/mol
CAS Number: 2052128-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR