1-Bromo-3-chloro-4-(methylthio)benzene 95%

Supplier: Ambeed
Warning

A714247-1G A714247-250MG
76651-426EA 149 USD
76651-426 76651-428
1-Bromo-3-chloro-4-(methylthio)benzene 95%
1-Bromo-3-chloro-4-(methylthio)benzene

Precaución: for Research Use Only

Formula: C₇H₆BrClS
MW: 237.5445 g/mol
MDL Number: MFCD22543648
CAS Number: 101084-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR