1,3-Dibromo-5-butoxybenzene 95%

Supplier: Ambeed
Warning

A713789-1G A713789-250MG A713789-5G
76651-412EA 201.58 USD
76651-412 76651-414 76651-416
1,3-Dibromo-5-butoxybenzene 95%
1,3-Dibromo-5-butoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂Br₂O
MW: 308.0097 g/mol
MDL Number: MFCD29043831
CAS Number: 896745-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR