S-Methyl-S-(4-chlorophenyl) sulfoximine 95%

Supplier: Ambeed
Warning

A713527-100MG A713527-250MG
76651-408EA 37 USD
76651-408 76651-410
S-Methyl-S-(4-chlorophenyl) sulfoximine 95%
S-Methyl-S-(4-chlorophenyl) sulfoximine

Precaución: for Research Use Only

Formula: C₇H₈ClNOS
MDL Number: MFCD00173771
CAS Number: 22132-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR