1,4-Dioxa-10-azadispiro[4.2.4.2]tetradecan-11-one 95%

Supplier: Ambeed
Warning

A713387-250MG A713387-5G A713387-1G
76651-404EA 60.48 USD
76651-404 76651-406 77617-936
1,4-Dioxa-10-azadispiro[4.2.4.2]tetradecan-11-one 95%
1,4-Dioxa-10-azadispiro[4.2.4.2]tetradecan-11-one

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₃
MW: 211.25 g/mol
MDL Number: MFCD24566289
CAS Number: 914780-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR