6-Bromo-3,3-dimethyl-1H,2H,3H-pyrrolo[3,2-b]pyridin-2-one 98%

Supplier: Ambeed
Warning

A712961-100MG A712961-1G A712961-250MG
76651-396EA 49.87 USD
76651-396 76651-398 77622-536
6-Bromo-3,3-dimethyl-1H,2H,3H-pyrrolo[3,2-b]pyridin-2-one 98%
6-Bromo-3,3-dimethyl-1H,2H,3H-pyrrolo[3,2-b]pyridin-2-one

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O
MW: 241.0846 g/mol
MDL Number: MFCD23097462
CAS Number: 1190862-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR