3-Fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 95%
Supplier: Ambeed
|
Warning
|
A711604-100MG
A711604-1G
A711604-250MG
A711604-5G
76651-362EA
37.14
USD
76651-362
76651-364
76651-366
76651-368
3-Fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 95%
3-Fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₃H₁₆BFO₃ MW: 250.0800 g/mol |
MDL Number:
MFCD18383509 CAS Number: 1246633-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...