2-Cyano-3-(4-hydroxy-3,5-diisopropylphenyl)acrylamide 98%

Supplier: Ambeed
Warning

A709920-10MG A709920-1MG A709920-50MG A709920-5MG
76651-340EA 41 USD
76651-340 76651-342 76651-344 76651-346
2-Cyano-3-(4-hydroxy-3,5-diisopropylphenyl)acrylamide 98%
2-Cyano-3-(4-hydroxy-3,5-diisopropylphenyl)acrylamide

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₂
MDL Number: MFCD31544351
CAS Number: 106392-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR