(S)-2-Amino-4-((4-chlorobenzyl)amino)-1-(piperidin-1-yl)butan-1-one dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: UAMC00039 2HCl

A709631-10MG A709631-1MG A709631-25MG A709631-50MG A709631-5MG
76651-330EA 66.84 USD
76651-330 76651-332 76651-334 76651-336 76651-338
(S)-2-Amino-4-((4-chlorobenzyl)amino)-1-(piperidin-1-yl)butan-1-one dihydrochloride 98%
(S)-2-Amino-4-((4-chlorobenzyl)amino)-1-(piperidin-1-yl)butan-1-one dihydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₆Cl₃N₃O
MW: 382.7561 g/mol
MDL Number: MFCD27937768
CAS Number: 697797-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR