4,4,4-Trifluoro-1-(thiophen-2-yl)-2-(2-(m-tolyl)hydrazono)butane-1,3-dione 98%

Supplier: Ambeed
Warning

A709349-10MG A709349-1MG A709349-25MG A709349-50MG A709349-5MG
76651-320EA 64 USD
76651-320 76651-322 76651-324 76651-326 76651-328
4,4,4-Trifluoro-1-(thiophen-2-yl)-2-(2-(m-tolyl)hydrazono)butane-1,3-dione 98%
4,4,4-Trifluoro-1-(thiophen-2-yl)-2-(2-(m-tolyl)hydrazono)butane-1,3-dione

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃N₂O₂S
MDL Number: MFCD00096392
CAS Number: 263890-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR