Mycro-3 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-(7-(chlorodifluoromethyl)-5-(furan-2-yl)pyrazolo[1,5-a]pyrimidine-2-carboxamido)-1-phenyl-1H-pyrazole-4-carboxylate, Mycro 3

A708851-10MG A708851-1MG A708851-25MG A708851-50MG A708851-5MG
76651-298EA 50.93 USD
76651-298 76651-300 76651-302 76651-304 76651-306
Mycro-3 98%
Mycro-3

Precaución: for Research Use Only

Formula: C₂₄H₁₇ClF₂N₆O₄
MW: 526.8794 g/mol
MDL Number: MFCD03409284
CAS Number: 944547-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR