Succinobucol 98%

Supplier: Ambeed
Warning

A708788-10MG A708788-1MG A708788-25MG A708788-50MG A708788-5MG A708788-100MG
76651-288EA 43 USD
76651-288 76651-290 76651-292 76651-294 76651-296 77783-698
Succinobucol 98%
Succinobucol

Precaución: for Research Use Only

Formula: C₃₅H₅₂O₅S₂
MDL Number: MFCD08690469
CAS Number: 216167-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR