Tropifexor 98%

Supplier: Ambeed
Warning

Synonyms: rel-2-((1R,3r,5S)-3-((5-cyclopropyl-3-(2-(trifluoromethoxy)phenyl)isoxazol-4-yl)methoxy)-8-azabicyclo[3.2.1]octan-8-yl)-4-fluorobenzo[d]thiazole-6-carboxylic acid

A708662-10MG A708662-1MG A708662-50MG A708662-5MG
76651-272EA 234 USD
76651-272 76651-274 76651-276 76651-278
Tropifexor 98%
Tropifexor

Precaución: for Research Use Only

Formula: C₂₉H₂₅F₄N₃O₅S
MDL Number: MFCD31693060
CAS Number: 1383816-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR