2-(2-(Prop-2-yn-1-yloxy)ethoxy)acetic acid 95%

Supplier: Ambeed
Danger

Synonyms: Propynyl-PEG1-Ac

A706247-100MG A706247-1G A706247-250MG A706247-5G
76651-260EA 46 USD
76651-260 76651-262 76651-264 77598-728
2-(2-(Prop-2-yn-1-yloxy)ethoxy)acetic acid 95%
2-(2-(Prop-2-yn-1-yloxy)ethoxy)acetic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
MW: 158.1519 g/mol
MDL Number: MFCD30469070
CAS Number: 944561-45-9
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR