HO-PEG3-CH2COOH 95%

Supplier: Ambeed
Warning

Synonyms: PEG3-O-CH2COOH

A705669-100MG A705669-1G A705669-250MG
76651-254EA 40 USD
76651-254 76651-256 76651-258
HO-PEG3-CH2COOH 95%
HO-PEG3-CH2COOH

Precaución: for Research Use Only

Formula: C₈H₁₆O₆
MDL Number: MFCD24479936
CAS Number: 51951-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR