3-Chloro-4-iodobenzonitrile 97%

Supplier: Ambeed
Danger

A704603-100MG A704603-1G A704603-250MG A704603-5G
76651-246EA 10 USD
76651-246 76651-248 76651-250 76651-252
3-Chloro-4-iodobenzonitrile 97%
3-Chloro-4-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClIN
MDL Number: MFCD18393347
CAS Number: 1261686-46-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR