3,6,9,12,15-Pentaoxaoctadec-17-yn-1-ol 95%
Supplier: Ambeed
|
Warning
|
A703033-100MG
A703033-250MG
A703033-5G
A703033-1G
76651-226EA
14
USD
76651-226
76651-228
76651-230
77598-726
3,6,9,12,15-Pentaoxaoctadec-17-yn-1-ol 95%
3,6,9,12,15-Pentaoxaoctadec-17-yn-1-ol
Precaución: for Research Use Only
Formula:
C₁₃H₂₄O₆ |
MDL Number:
MFCD27635181 CAS Number: 1036204-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...