4,6-Dichloropyridazine-3-carbonitrile 95%

Supplier: Ambeed
Warning

A702715-100MG A702715-250MG A702715-1G
76651-222EA 51.99 USD
76651-222 76651-224 77573-272
4,6-Dichloropyridazine-3-carbonitrile 95%
4,6-Dichloropyridazine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₅HCl₂N₃
MW: 173.9875 g/mol
MDL Number: MFCD31620743
CAS Number: 35857-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR