YU238259 97%

Supplier: Ambeed
Warning

Synonyms: N-(2-(5-chloropyridin-2-yl)ethyl)-4-(((4-methoxyphenyl)sulphonamido)methyl)benzamide

A699629-10MG A699629-1MG A699629-25MG A699629-5MG
76651-196EA 27.59 USD
76651-196 76651-198 76651-200 76651-202
YU238259 97%
YU238259

Precaución: for Research Use Only

Formula: C₂₂H₂₂ClN₃O₄S
MW: 459.94 g/mol
MDL Number: MFCD30532769
CAS Number: 1943733-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR