tert-Butyl 9-aminononanoate 98%

Supplier: Ambeed
Danger

A699139-1G A699139-250MG
76651-188EA 65 USD
76651-188 77602-770
tert-Butyl 9-aminononanoate 98%
tert-Butyl 9-aminononanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₇NO₂
MW: 229.3590 g/mol
CAS Number: 134857-22-0
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR