(2-Chloro-5-fluoro-3-methylphenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-fluoro-3-methylphenylboronic acid

A698720-100MG A698720-1G A698720-250MG A698720-5G
76651-176EA 45.62 USD
76651-176 76651-178 76651-180 76651-182
(2-Chloro-5-fluoro-3-methylphenyl)boronic acid 95%
(2-Chloro-5-fluoro-3-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₂
MW: 188.3917 g/mol
MDL Number: MFCD03788415
CAS Number: 2121511-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR