Methyl 4-(bromomethyl)-2-chlorobenzoate 97%
Supplier: Ambeed
|
Danger
|
A697964-100MG
A697964-1G
A697964-250MG
A697964-5G
76651-166EA
21
USD
76651-166
76651-168
76651-170
76651-172
Methyl 4-(bromomethyl)-2-chlorobenzoate 97%
Methyl 4-(bromomethyl)-2-chlorobenzoate
Precaución: for Research Use Only
Formula:
C₉H₈BrClO₂ MW: 263.5156 g/mol |
MDL Number:
MFCD18074272 CAS Number: 143572-60-5 UN: 3261 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...