(S)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-aminohexanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Lys-OtBu·HCl
A696236-250MG
A696236-1G
A696236-25G
A696236-5G
76651-158EA
27
USD
76651-158
77552-108
77552-110
77552-112
(S)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-aminohexanoate hydrochloride 97%
(S)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-aminohexanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₂₅H₃₃ClN₂O₄ MW: 460.99 g/mol |
MDL Number:
MFCD32066818 CAS Number: 2413365-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...