2-(4-Ethylphenyl)-2-oxoacetic acid 95%

Supplier: Ambeed
Warning

A696192-100MG A696192-1G
76651-154EA 42.44 USD
76651-154 76651-156
2-(4-Ethylphenyl)-2-oxoacetic acid 95%
2-(4-Ethylphenyl)-2-oxoacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.1800 g/mol
MDL Number: MFCD11179648
CAS Number: 7099-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR