Cyclooct-4-enecarboxylic acid 95%

Supplier: Ambeed
Warning

A693064-100MG A693064-250MG
76651-128EA 214.31 USD
76651-128 76651-130
Cyclooct-4-enecarboxylic acid 95%
Cyclooct-4-enecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₄O₂
MW: 154.2063 g/mol
MDL Number: MFCD00019726
CAS Number: 4103-10-0
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR