26-Amino-3,6,9,12,15,18,21,24-octaoxahexacosan-1-ol 95%

Supplier: Ambeed
Warning

Synonyms: Amino-PEG9-alcohol

A692648-100MG A692648-250MG
76651-124EA 80.63 USD
76651-124 76651-126
26-Amino-3,6,9,12,15,18,21,24-octaoxahexacosan-1-ol 95%
26-Amino-3,6,9,12,15,18,21,24-octaoxahexacosan-1-ol

Precaución: for Research Use Only

Formula: C₁₈H₃₉NO₉
MW: 413.5036 g/mol
MDL Number: MFCD31656979
CAS Number: 15332-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR