2-Benzyl-3-(4-chlorophenyl)-5-(4-nitrophenyl)pyrazolo[1,5-a]pyrimidin-7(4H)-one 98%

Supplier: Ambeed
Warning

Synonyms: NAV-2729

A691659-10MG A691659-1MG A691659-25MG A691659-50MG A691659-5MG
76651-114EA 97 USD
76651-114 76651-116 76651-118 76651-120 76651-122
2-Benzyl-3-(4-chlorophenyl)-5-(4-nitrophenyl)pyrazolo[1,5-a]pyrimidin-7(4H)-one 98%
2-Benzyl-3-(4-chlorophenyl)-5-(4-nitrophenyl)pyrazolo[1,5-a]pyrimidin-7(4H)-one

Precaución: for Research Use Only

Formula: C₂₅H₁₇ClN₄O₃
MDL Number: MFCD01455625
CAS Number: 419547-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR