3-(Bromomethyl)-5-fluoropyridine hydrobromide 95%

Supplier: Ambeed
Danger

A691086-100MG A691086-250MG A691086-1G
76651-110EA 161.26 USD
76651-110 76651-112 77617-864
3-(Bromomethyl)-5-fluoropyridine hydrobromide 95%
3-(Bromomethyl)-5-fluoropyridine hydrobromide

Precaución: for Research Use Only

Formula: C₆H₆Br₂FN
MW: 270.9249 g/mol
MDL Number: MFCD26407887
CAS Number: 1256561-65-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR