(S)-N,N-Dimethyl-3-pyrrolidinecarboxamide hydrochloride 95%

Supplier: Ambeed
Warning

A690256-100MG A690256-250MG
76651-100EA 171.87 USD
76651-100 76651-102
(S)-N,N-Dimethyl-3-pyrrolidinecarboxamide hydrochloride 95%
(S)-N,N-Dimethyl-3-pyrrolidinecarboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅ClN₂O
MW: 178.6598 g/mol
MDL Number: MFCD20487825
CAS Number: 1315593-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR