6-Bromopyrazolo[1,5-a]pyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A689114-100MG A689114-1G A689114-250MG
76651-082EA 102.91 USD
76651-082 76651-084 76651-086
6-Bromopyrazolo[1,5-a]pyridine-3-carbonitrile 97%
6-Bromopyrazolo[1,5-a]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrN₃
MW: 222.0415 g/mol
MDL Number: MFCD31728303
CAS Number: 2092835-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR