Methyl 3-(2-((5-(3,5-dimethoxyphenyl)-1,3,4-oxadiazol-2-yl)thio)acetamido)benzoate 97%

Supplier: Ambeed
Warning

A1349702-100MG A1349702-250MG
76658-126EA 240.83 USD
76658-126 76658-128
Methyl 3-(2-((5-(3,5-dimethoxyphenyl)-1,3,4-oxadiazol-2-yl)thio)acetamido)benzoate 97%
Methyl 3-(2-((5-(3,5-dimethoxyphenyl)-1,3,4-oxadiazol-2-yl)thio)acetamido)benzoate

Precaución: for Research Use Only

Formula: C₂₀H₁₉N₃O₆S
MW: 429.4464 g/mol
MDL Number: MFCD05842283
CAS Number: 838816-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR