4-Cyclohexyl-2,6-dimethoxyphenol 97%

Supplier: Ambeed
Warning

A1349690-100MG A1349690-250MG
76658-122EA 181 USD
76658-122 76658-124
4-Cyclohexyl-2,6-dimethoxyphenol 97%
4-Cyclohexyl-2,6-dimethoxyphenol

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₃
MDL Number: MFCD33022389
CAS Number: 2682114-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR