1-octyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide 98%

Supplier: Ambeed
Warning

A687763-1G A687763-25G A687763-5G
76651-060EA 18.04 USD
76651-060 76651-062 76651-064
1-octyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide 98%
1-octyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₆H₂₃F₆N₃O₂
MW: 403.3632 g/mol
MDL Number: MFCD19443502
CAS Number: 862731-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR