(4,4-Difluorocyclohexyl)hydrazine hydrochloride 97%

Supplier: Ambeed
Warning

A687708-100MG A687708-250MG A687708-1G
76651-056EA 60 USD
76651-056 76651-058 77783-651
(4,4-Difluorocyclohexyl)hydrazine hydrochloride 97%
(4,4-Difluorocyclohexyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃ClF₂N₂
MDL Number: MFCD30802276
CAS Number: 1548590-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR