10-((2,5-Dioxopyrrolidin-1-yl)oxy)-10-oxodecanoic acid 95%

Supplier: Ambeed
Warning

A687135-100MG A687135-1G A687135-250MG
76651-050EA 123 USD
76651-050 76651-052 76651-054
10-((2,5-Dioxopyrrolidin-1-yl)oxy)-10-oxodecanoic acid 95%
10-((2,5-Dioxopyrrolidin-1-yl)oxy)-10-oxodecanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₆
MDL Number: MFCD31698736
CAS Number: 159350-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR