Ethyl 3-Methylpyrazole-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-methyl-1H-pyrazole-5-carboxylate

A686780-100G A686780-10G A686780-25G A686780-5G
76651-042EA 227.04 USD
76651-042 76651-044 76651-046 76651-048
Ethyl 3-Methylpyrazole-5-carboxylate 95%
Ethyl 3-Methylpyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.165 g/mol
MDL Number: MFCD00052514
CAS Number: 1226862-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR